VWR® Drosophila Refrigerated Peltier Incubator, High Efficiency
Proveedor: VWR, part of Avantor
VRI20PD VRI20PD-2
89510-750EA 251251.05 MXN
89510-750 89510-752
VWR® Drosophila Refrigerated Peltier Incubator, High Efficiency
Incubadoras Incubadoras refrigeradas
Using innovative peltier cooling technology, the VWR drosphila incubators offer a 78% reduction in energy consumption over traditional compressor based refrigerated incubators.
  • Robust design
  • Whisper-quiet operation
  • Over-temperature thermostat and alarm
These incubators are equipped with LED Lighting and Day/Night Controls to create ideal conditions for fruit flies
Independent over-temperature thermostat and alarm.
The robust cabinet design includes a mechanical convection system to provide improved temperature uniformity, digital temperature controller with display, and over temperature limit control. The compressor-free design creates a whisper-quiet operation.
Certificaciones: TUV, CE, UL, CSA.
Información para pedidos: Three-year parts and labor warranty (domestic).