Orion™ Nitrate Electrodes, Thermo Scientific
Proveedor: Thermo Scientific
Orion™
930701 930702 970701 9707BNWP 9707BNWP
21000-046EA 28620.64 MXN
34112-581 21000-046 34112-586 34118-106 CA21000-046
Orion™ Nitrate Electrodes, Thermo Scientific
Electrodos Electrodos selectivos de iones (ISE)
Measure nitrate in aqueous solutions simply, accurately, and economically with Orion™ Nitrate Electrodes.
  • Drift-free potentials
  • Free-flowing Sure-Flow junction
A separate reference electrode is unnecessary, making these electrodes convenient to use with small sample volumes.