SP Bel-Art Wash Bottle, 2-Color, Safety-Labeled, Wide-Mouth, LDPE, Bel-Art Products, a part of SP
Proveedor: Bel-Art
SCIENCEWARE®
F11646-0639 F11646-0623 F11646-6250 F11646-0621 F11646-0628 F11646-0620 F11646-0611 F11646-1132 F11646-0640 F11646-0624 F11646-0612 F11646-2432 F11646-0617 F11646-0614 F11646-0622 F11646-0638 F11646-3832 F11646-6250 F11646-3739 F11646-0631 F11646-0627 F11646-0050 F11646-6032 F11646-2232
89141-034CS 2392.61 MXN
89141-012 89141-034 89141-014 70690-068 490015-562 89141-016 70690-062 70690-073 70690-051 89501-664 70690-093 70690-285 70690-066 70690-074 70690-086 70690-064 89501-662 74900-008 74900-006 74900-004 74900-012 74900-002 70680-363 16649-923
SP Bel-Art Wash Bottle, 2-Color, Safety-Labeled, Wide-Mouth, LDPE, Bel-Art Products, a part of SP
Frascos Frascos de lavado
Bel-Art’s Safety Labeled wash bottles for dispensing your most common solvents.
  • Polypropylene closure with leakproof design
  • Easy to read
  • LDPE solvent-resistant
Printed bottles include: Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making the workplace a safer place.
Certificaciones: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Información para pedidos: Assortment includes one each of Acetone, Ethanol, Isopropanol, Methanol, Toluene, and Water. Use only with solvents compatible with LDPE.