SP Bel-Art Microcentrifuge Tube Ice Rack/Tray and Cover, Bel-Art Products, a part of SP
Catalog # 47751-770
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacity:0.5 L (17 oz.)
- Description:Rack
- Dimensions:23.2L x 13.3W x 7.3H cm (9⅛ x 5¼ x 2⅞")
- Cat. no.:47751-770
- Supplier No.:189050001
Specifications
About this item
This tray with rack insert can accommodate up to fifty 1.5 mL tubes (or other tubes with a diameter less than 12 mm) for large volume benchtop processing.
- Sturdy polypropylene construction
- Autoclavable
- Stackable
- Designed for long life and heavy usage
It features a deep tray to hold more ice and water for extended use, a flat perimeter rim for labeling tape, and it can be safely stacked.
To protect tubes during transport or storage while also maintaining stackability, an optional polypropylene cover is available.
Designed for long life and heavy usage, all parts are molded of sturdy polypropylene and are steam autoclavable at 121 °C (250 °F).