Orion™ ORP Standards for Redox/ORP Electrodes, Thermo Scientific
Proveedor: Thermo Scientific
967961 967901 967901 967961
46610-338EA 1623.05 MXN
46610-336 CA46610-337 46610-338 CA46610-338
Orion™ ORP Standards for Redox/ORP Electrodes, Thermo Scientific
Normativas Estándar Redox/ORP
Used as relative mV standard to calibrate the offset of sample.
Stable for up to a year if unopened and can be used directly from the bottle with no mixing or diluting.
ORP standard does not contain hazardous chemicals.