Chemical Glass Beads, Propper Manufacturing
Proveedor: Propper
03000900 03000200 03001000 03000800 03000600 03000400
48300-446PK 1254.24 MXN
48300-446 48300-448 48300-437 48300-440 48300-454 48300-433
Chemical Glass Beads, Propper Manufacturing
Perlas Microesferas de vidrio
Propper solid chemical beads are designed to reduce bumping during boiling, refluxing, and other distilling operations.
  • Designed to reduce bumping during distillation
  • Soda-lime glass beads resistant to extreme temperatures
  • Available in a variety of sizes
Propper glass beads are made of soda-lime glass and are available in diameters from 1 to 6 mm, sizing held close to tolerances.
Designed for use wherever a large surface area of glass is desirable (packed columns for absorption, distillation, etc.). Beads are supplied 0.45 Kg (1 lb.) per box.