AccuVac® Ampule Snapper, Hach
Proveedor: Hach
AccuVac®
2405200 2405200 24052-00
89500-730EA 1100 MXN
CA24052-00 CA89500-730 89500-730
AccuVac® Ampule Snapper, Hach
Ampoule Openers
These snappers are specially used for opening ampule immersed in sample.
Avoid sample contact and mainly used with colorimeters and spectrophotometers.