Stomacher® Bags for Stomacher® 400 Lab Blender Series, Seward
Proveedor: Seward
Stomacher®
030020162 030020278 030020219 030020251 BA6041 030020235
58976-930PK 8633.72 MXN
58976-930 58976-921 58976-955 58976-957 58976-925 58976-959
Stomacher® Bags for Stomacher® 400 Lab Blender Series, Seward
Bolsas Bolsas para homogeneizadores
The Stomacher® Bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending (processes requiring more than 15 minutes on the machine) and small samples for life science applications.
  • Made from high quality food grade virgin LDPE granule extruded to 65 µm film thickness
  • Withstand extensive blending without tears or perforations
  • Low cost option for sample preparation with no risk of cross contamination between samples
  • Irradiated sterile
Bags for the Stomacher® 400 have a capacity of 80 to 400 ml. Compatible with all lab blenders and homogenizers.
Información para pedidos: Irradiation certificates are available on request.