BODTrak II Respirometric BOD Apparatus, Hach
Proveedor: Hach
2952400 2952400
89500-782EA 212900 MXN
89500-782 CA11024-820
BODTrak II Respirometric BOD Apparatus, Hach
DBO, Demanda biológica de oxígeno
  • BODTrak II decreases total test time
  • Faster than Dilution Method
  • BOD results are easy to monitor
  • Standalone operation
  • DO probes and titrations are eliminated
Simplify BOD analysis with the BODTrak™II Respirometric BOD Apparatus. Results comparable to the dilution method can be achieved in only two to three days. Use these results for process control or as a complement to the dilution method (not approved by USEPA for NPDES reporting purposes). The BODTrak II is a re-engineered version of the original Hach BODTrak. Improvements to the old system include greaseless bottle seals, a larger graphical display, improved stirring and temperature control, a smaller footprint, improved procedures with expanded options, an auto-switching power supply, and bottle fences to prevent tipping of bottles.
Información para pedidos: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.