Orion™ Electrode Adapter Cables, Thermo Scientific
Proveedor: Thermo Scientific
Orion™
090036 91CBNC 91USRF 91CLM0 090034
34104-743EA 2234.62 MXN
34104-751 23198-218 23198-308 34104-743 34104-742
Orion™ Electrode Adapter Cables, Thermo Scientific
Cabos
Can convert an electrode with one connector to the required connector of another meter by using this cable.
For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2 mm reference pins, or just a stripped lead inputs. Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connector.