SP Bel-Art Wash Bottles, Wide Mouth, Bel-Art Products, a part of SP
Proveedor: Bel-Art
SCIENCEWARE®
F11626-0500 F11626-0250 F11620-1000 F11620-0125 F11628-0500 F11620-0250 F11625-1000 F11628-0250 F11627-1000 F11626-1000 F11627-0250 F11628-1000 F11625-0250 F11620-0500 F11627-0500
16650-253CS 3618.38 MXN
16650-014 70690-056 70690-078 16650-008 16650-018 16650-016 70690-082 10033-462 11215-300 16650-253 16650-275 10844-026 16650-022 16650-020 47748-154
SP Bel-Art Wash Bottles, Wide Mouth, Bel-Art Products, a part of SP
Frascos Frascos de lavado
Low-density polyethylene wash bottles have color-coded polypropylene closures for easy identification and wide mouths for easy filling.
  • Color-coded closures for quick identification
  • Wide-mouthed design
  • LDPE construction
Choose from Natural, Red, Yellow, Blue, and Green caps in 250, 500, or 1,000 ml sizes. Natural caps are also available in 125 ml size.