VWR® Basic Refrigerators with Glass Doors and Natural Refrigerant
Proveedor: VWR
VWR®
GDM72WHTSS GDM47HCVWRWHT GDM23HC GDM41HCVWRWHT GDM49WHTSS GDM49HC GDM72HC GDM23WHTSS
75871-650EA 172392.3 MXN
75871-648 10791-618 10018-864 10018-866 75871-650 10791-626 10018-860 10791-624
VWR® Basic Refrigerators with Glass Doors and Natural Refrigerant
Refrigeradores
Swinging or sliding doors. These upright laboratory refrigerators maintain an average temperature of 3 – 5 °C (37 – 41 °F).
  • All units feature a natural refrigerant that is environmentally friendly
  • R290 hydrocarbon refrigerant emits zero ozone depletion potential
  • Units are supplied standard with the LAE temperature control
  • Exterior digital display and alarm function
Units are equipped with low E, double-pane, thermal insulated glass doors for energy efficiency.
Note: Units do not come with castors. Castors can be ordered separately.
These refrigerators are supplied standard with four, eight, or twelve adjustable, heavy-duty, PVC-coated full white wire shelves.
Certificaciones: C-UL US listed. NSF certified. Natural Refrigerant.
Información para pedidos: Additional full white wire shelves can be ordered seperately for any VWR® double glass door refrigerators. Optional chart recorder, alarm, and thermometer accessories require factory installation.
Warranty: Refrigerators come with a three-year parts and labor warranty, with an additional two-year warranty on for compressor parts.