NewClassic Level, TLE Series Precision Balances, METTLER TOLEDO®
30046476 30046473 30046473 30046476
10031-618EA 48014.74 MXN
CA10031-616 10031-618 10031-616 CA10031-618
NewClassic Level, TLE Series Precision Balances, METTLER TOLEDO®
TLE Series Precision balances offer effortless cleaning, comfortable daily use, and environmental friendliness. They are the perfect solution for classroom applications.
  • Capacities from 320 to 3200 g
  • Readabilities from 0.2 mg
  • Meets all GLP and ISO 9001 Requirements
NewClassic TLE Balances support teaching and learning
These robust balances are designed to withstand intensive use and to provide years of reliable results.
Teaching Lab products are specifically designed for the needs of the classroom – to make teaching and learning as easy as possible for both teachers and students. Daily weighing tasks are easily executed thanks to the five clear and intuitive function keys. Applications can be accessed at the touch of a button and the large numbers on the brilliant backlit display are easy to read.
Certificaciones: METTLER TOLEDO® provides ISO 9001 manufacturing, nationwide ISO 9001 field service, ISO 9001 quality certification, and ISO 14001 environmental certification. UL listed. CSA certified.
Información para pedidos: TLE Series Precision balances are packaged with appropriate power supply.