Order Entry
VWR® Gravity Convection Ovens
VWR® Gravity Convection Ovens
Supplier:  VWR International
Certificates
undefined
VWR® Gravity Convection Ovens
Supplier:  VWR International
Specifications
  • Catalog No:
  • 89511-408
  • 89511-406
  • 89511-404
  • Temperature fluctuation:
  • ± 0.4 °C at 150°C
  • ± 0.4 °C at 150°C
  • ± 0.4 °C at 150°C
  • Temperature range:
  • 50 - 250°C
  • 50 - 250°C
  • 50 - 250°C
  • Temperature setting accuracy:
  • 0.1°C
  • 0.1°C
  • 0.1°C
  • Electrical:
  • 120V
  • 120V
  • 120V
Specifications
Related Information
Gravity Ovens
Product Information
  • Catalog #
    Volume
    Exterior Dimensions
    Heat-Up Time
    Recovery Rate
    Temperature Uniformity
    Shipping Weight
    Availability
    Price
  • 89511-404
    65 L (2.3 cu. ft.)
    22D x 21W x 28H"
    25 min. to 150 °C
    6 min to 150 °C
    ±4 °C at 150 °C (based on 27 sensors)
    121.25 lbs.
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $74,088.02
    $74,088.02
    Each (1UN)
       $74,088.02
    Specifications:
    CSA Certified
    Product available on GSA Advantage®
    More
    Accessories
    Specifications
    Environmentally Preferable
    Temperature fluctuation
    ± 0.4 °C at 150°C
    Temperature range
    50 - 250°C
    Temperature setting accuracy
    0.1°C
    Electrical
    120V
    CSA Certified
    Product available on GSA Advantage®
  • 89511-406
    105 L (3.7 cu. ft.)
    22D x 25W x 32H"
    24 min. to 150 °C
    8 min to 150 °C
    ±4.5 °C at 150 °C (based on 27 sensors)
    143.3 lbs.
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $86,880.19
    $86,880.19
    Each (1UN)
       $86,880.19
    Specifications:
    CSA Certified
    Product available on GSA Advantage®
    More
    Accessories
    Specifications
    Environmentally Preferable
    Temperature fluctuation
    ± 0.4 °C at 150°C
    Temperature range
    50 - 250°C
    Temperature setting accuracy
    0.1°C
    Electrical
    120V
    CSA Certified
    Product available on GSA Advantage®
  • 89511-408
    176 L (6.2 cu. ft.)
    29D x 25W x 36H"
    28 min. to 150 °C
    8 min to 150 °C
    ±5 °C at 150 °C (based on 27 sensors)
    189.6 lbs.
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $127,122.25
    $127,122.25
    Each (1UN)
       $127,122.25
    Specifications:
    CSA Certified
    More
    Accessories
    Specifications
    Environmentally Preferable
    Temperature fluctuation
    ± 0.4 °C at 150°C
    Temperature range
    50 - 250°C
    Temperature setting accuracy
    0.1°C
    Electrical
    120V
    CSA Certified
Frequently Bought Together