/
Mexico
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
AP-300 Polarimeters, ATAGO®
97015-538
:
ATAGO USA INC.
AP-300 Polarimeters, ATAGO®
97015-538
:
ATAGO USA INC.
:
5291
Model:
AP-300
Temperature Control:
18 to 30 °C
Cat. no.:
97015-538
Weight:
14.4 kg
Includes:
Polarimeter Only
Supplier No.:
5291