Rapidly dispense up to 45 times in succession without refilling.
- Positive displacement design ensures accurate results, even for viscous, volatile, or other problematic liquids
- Lightweight, ergonomic handle and wide finger rest minimize operator hand strain
- Operation is simple—just attach the appropriate tip size, fill and prime the tip, and dial the selected volume
- Volume is easy to set, with the aid of the convenient volume-setting chart located on the handle
The Finnpipette™ Stepper Pipettor uses Thermo Scientific Finntip™ Stepper tips, plunger-style tips available in nonsterile or sterile (individually wrapped) formats. This tip comes in seven sizes from 0.5 to 50 ml, each with five different delivery volumes, for an overall dispensing range of 10 to 5000 μl. Only the disposable tip comes in contact with the sample, eliminating the risk of contaminating the pipetter body.
The Thermo Scientific™ Finnpipette™ Stepper Pipetter is a lightweight, easy to use repeater pipetter ergonomically designed for one-handed dispensing. Operating on the positive displacement principle, the Finnpipette Stepper is ideally suited for work with aggressive and viscous liquids.
Información para pedidos: Purchase includes one nonsterile adapter that is necessary for use with 25 or 50 ml tips.