Latex Free Aneroid Sphygmomanometer
Catalog # 470017-351
Supplier: HEALTH SMART SE
Specifications
- Description:Latex Free Aneroid Sphygmomanometer
- Cat. no.:470017-351
- Supplier No.:09-149-011
Specifications
About this item
This economical sphygmomanometer measures blood pressure with a standard-size, Velcro arm cuff and easy to read dial. Accurate to +/- 3mm Hg, For easy access it stores in a handy zippered case.