Spectroquant® NOVA 60A Photometer, MilliporeSigma
Proveedor: MilliporeSigma
1.09752.0001 1.09752.0001
EM1.09752.0001EA 252346.8 MXN
CA1.09752.0001 EM1.09752.0001
Spectroquant® NOVA 60A Photometer, MilliporeSigma
Espectrofotómetros y Fotómetros Espectrofotómetros UV/Vis
The Spectroquant® NOVA 60 A are suitable for the routine analysis of all water types and capable of measuring all the ready-to-use cell tests as well as reagent tests.
  • Barcode reading of all Spectroquant® tests
  • Automatic selection of method and presentation of result
  • Integrated QA/QC checks - Three modes, checking instrument, the method and sample volume
  • Absorbance, concentration, transmission measuring modes
Designed for some critical tests, measurement to ppb levels, such as cyanide.