Spectroquant® Nitrogen (total) Cell Test Method: photometric, DMP 10 - 150 mg/L N
Proveedor: MilliporeSigma
Spectroquant®
1.14763.0001
EM1.14763.0001CS
61070.26
MXN
EM1.14763.0001
Spectroquant® Nitrogen (total) Cell Test Method: photometric, DMP 10 - 150 mg/L N
Kits para test medioambiente