Stainless Steel Scoop, Wheaton, DWK Life Sciences
Proveedor: DWK Life Sciences
885540
66174-540CS 4116.15 MXN
66174-540
Stainless Steel Scoop, Wheaton, DWK Life Sciences
Palas
This scoop is made from 302 stainless steel and is ideal for sampling powders, slurries, soils and more