Esta buscando: Bombas de vació
19  results were found
(IVA no incluido)
Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.
SearchResultCount:"19"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Numero del catalogo: (470121-430)
Proveedor: VWR International
Descripción: Efficient combination of a pump and a vacuum plate.
Numero del catalogo: (470149-508)
Proveedor: VWR International
Descripción: Sturdy Hand Pump Features Hose Connectors For Vacuum And Pressure
Numero del catalogo: (16210-206)
Proveedor: Thermo Fisher Scientific
Descripción: Obtain full blank port pressure with only two strokes when using the Thermo Scientific™ Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge, a...
Numero del catalogo: (26671-484)
Proveedor: LABCONCO CORPORATION MX
Descripción: Rotary Vane Direct Drive and Hybrid Vacuum Pumps provide a reliable vacuum source for a variety of laboratory products including FreeZone Freeze Dry S...
Proveedor: Thermo Fisher Scientific
Descripción: These durable pumps easily and quickly attain and hold vacuum of 635Torr (25" Hg)
Numero del catalogo: (89097-320)
Proveedor: ACE GLASS, INC.
Descripción: Two-stage, chemically resistant diaphragm type pump
Product available on GSA Advantage® Small Business Enterprise
Numero del catalogo: (89492-594)
Proveedor: ANTYLIA SCIENTIFIC
Descripción: Unique cavity configuration minimizes dead space and air entrapment with unique cavity configuration.
Proveedor: WELCH VACUUM BY GARDNER DENVER
Descripción: Utilice estas resistentes bombas estancas al aceite para una amplia variedad de necesidades de vacío en el laboratorio
Product available on GSA Advantage®
Numero del catalogo: (470110-926)
Proveedor: WELCH VACUUM BY GARDNER DENVER SE
Descripción: Welch DuoSeal is a single-stage pump with metal vanes and motor mounted on steel base with rubber feet.
Product available on GSA Advantage®
Numero del catalogo: (54972-709)
Proveedor: WELCH VACUUM BY GARDNER DENVER
Descripción: Pumps are ideal for continuous-duty applications, including vacuum distillations, glove box transfer chamber, residue solvent removal, and resin degas...
Product available on GSA Advantage®
Numero del catalogo: (470145-338)
Proveedor: BEL-ART PRODUCTS SE
Descripción: Aids in the transfer of acids and reagents and virtually eliminates accidental bottle breakage and spills.
Proveedor: WELCH VACUUM BY GARDNER DENVER
Descripción: These oil-free pumps are ideal for basic lab operations.
Product available on GSA Advantage®
Numero del catalogo: (470313-372)
Proveedor: GSC INTERNATIONAL SE
Descripción: More economical than an electric vacuum pump.
Numero del catalogo: (EVR140-00-001)
Proveedor: EDWARDS VACUUM
Descripción: The Edwards RV series of oil sealed rotary vane pumps are unique in their capability to offer both high vacuum and high throughput performance, which ...
Numero del catalogo: (470006-924)
Proveedor: UNITED SCIENTIFIC SUPPLIES
Descripción: Dual use pump for create a vacuum or pressurizing.
Proveedor: WELCH VACUUM BY GARDNER DENVER
Descripción: These dry vacuum pumps, designed to meet chemical duty laboratory needs, are ideal for rotary evaporation, concentrators, evaporators, vacuum ovens, f...
CSA Certified Product available on GSA Advantage®
Inquire for Price
El stock para este material es limitado pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
Este producto está marcado como restringido y sólo pueden ser adquiridos por cuentas aprobado para el envío. Si necesita más ayuda, contacte con nosotros en el (+52 55) 5005-0100 Opción 1.
Documentación adicional puede ser necesaria para comprar este artículo. Un representante de VWR se comunicará con usted si es necesario.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
Este producto ya no está disponible. Las alternativas pueden estar disponibles mediante la búsqueda con el número de catálogo VWR enumerado anteriormente. Si necesita más ayuda, por favor llame a Servicio al Cliente de VWR al (+52 55) 5005-0100 Opción 1.
1 - 16 of 19
no targeter for Bottom