SP Bel-Art Sterileware® White Sterile Sampling Scoops, Bel-Art Products, a part of SP
Proveedor: Bel-Art
Sterileware®
H36902-0010 H36906-0010 H36904-0010 H36902-0000 H36904-0000 H36906-0000
89140-586PK 1025.55 MXN
89140-586 56924-503 56924-536 56924-569 89140-590 89140-594
SP Bel-Art Sterileware® White Sterile Sampling Scoops, Bel-Art Products, a part of SP
Palas
Sterileware® scoops are the perfect shape and size for taking samples, especially from wide-mouth containers.
  • Sterile
  • High-impact polystyrene plastic construction
  • White, one-piece design
  • Lot stamped for accurate lot tracing
  • Long handle improves reach and safety
Designed to hold various capacities, these one-piece sterile scoops are molded from high-impact polystyrene plastic, individually sealed and sterilized in a polyethylene bag, and ready for one-time use and disposal.
The 60 mL (2oz.) and 120 mL (4oz.) sampling scoops have a long handle that increases the distance between the hand and sample. When placed on a flat surface they remain upright and can be used as weighing boats.
Made from FDA-grade materials, they are lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization.