Incubators, Refrigerated with Peltier Technology, KT Series, BINDER
Proveedor: Binder
9020-0312 9020-0310 9020-0314
10031-394EA 8699.54 MXN
75838-570 10031-394 10823-814
Incubators, Refrigerated with Peltier Technology, KT Series, BINDER
Incubadoras Incubadoras refrigeradas
The KT refrigerated incubators with Peltier technology combine outstanding performance with environmental friendliness and provide sample protection.
These units feature the electronically controlled APT.line™ preheating chamber provide even horizontal airflow on both sides for homogenous temperature distribution along with long term stability. The units utilize Peltier refrigeration technology for finely controlled cooling from 4° to 100°C as well as a powerful conventional tubular heating system.
The disinfection routine is performed at 100°C in a high quality stainless steel inner chamber with easily removable elements and no permanent fixtures. The condensation-free inner chamber features an inner tempered safety glass door with a very tight seal and 2 point closure with 2.36” insulation for low heat dissipation.
Models feature a LCD screen with push button rotary knob and user-friendly menu. The variable programming options include 100 programs with 2 GB memory and a USB port for easy data transfer. Units have an Ethernet interface for use with APT-COM™ DataControlSystem communication software. Another feature is the independent temperature safety device class 3.1 with full over-temperature protection with visual and audible alarms. Options include key lock and password access protection to eliminate accidental or unauthorized access.
Información para pedidos: Units up to 115 liters ( 4.1 cubic feet) are stackable. All units come with a BINDER test confirmation. Units also come equipped with 2 stainless steel shelves.