Axygen® Disposable Reagent Reservoirs, Corning
Proveedor: Corning
Axygen®
RES-V-50-SI RES-2CV-25-SI RES-V-25-SI RES-V-100-S RES-V-50 RES-V-100 RES-V-100-SI RES-V-25-S RES-2CV-25 RES-V-50-S RES-V-25 RES-2CV-25-S
89108-002CS 2974.58 MXN
89108-004 89108-014 89108-002 89108-012 89108-022 89108-000 89108-010 89108-020 89108-008 89108-018 89108-006 89108-016
Axygen® Disposable Reagent Reservoirs, Corning
Depósitos de reactivos
These disposable reagent reservoirs are sterile and certified nonpyrogenic
They feature a sharp-V bottom that allows efficient extraction of the reagent. They also feature four pour-off corners and a stacking lug for easy separation