Low Intensity UV Lamp, Electron Microscopy Sciences
Proveedor: Electron Microscopy Sciences
72424 72423 72422 72425
100491-962EA 5449.28 MXN
100491-958 100491-962 100491-960 100491-964
Low Intensity UV Lamp, Electron Microscopy Sciences
Luces Lámparas de rayos UV
This 6-watt UV lamp meets most laboratory requirements; such as, bacterial identification, fluorchemistry, specimen staining, titration processes, photochemical reactions, aflatoxin detection, and curing UV cements. Constructed from Cyolac® plastic, with a longwave emission of 365 nm and an intensity at 6" (15.2 cm) of 750 µm/cm2. The bulb has a rated life of 6,000 hours. It can be used as a hand-held light or it may br mounted on a optional stand. Powered by 110V/60Hz or 220V/50Hz.