Septa, Thermo Scientific
Proveedor: Thermo Scientific
31303230 313G3230 313G3228 31303240 31303233-BP 31303211 313G3211 31303228 313G3240
10056-088EA 3139.5 MXN
10141-486 10056-070 10056-080 10056-090 10056-086 10056-084 10056-068 10056-088 10056-066
Septa, Thermo Scientific
Septos
Thermo Scientific™ BTO and TR-Green septa for GC instruments.
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.