VWR® Neoprene Crucible Holders
Proveedor: VWR
11648-286 11648-283 11648-284 11648-357 11648-281 11648-282 11648-285
89259-862CS 1869 MXN
89259-862 10200-148 89259-864 89259-860 89259-866 89259-868 89259-858
VWR® Neoprene Crucible Holders
Soportes Mangas protectoras
These neoprene crucible holders are tapered, collar-type holders.
  • Used alone or with adjacent sizes
They support funnels, crucibles, and other equipment in vacuum-type filtration assemblies. They feature a positive, but easy-to-release gas-tight seal.