VWR® Micro Spatulas, Double-Ended
Proveedor: Avantor
VWR®
11648-295 11648-297 11648-296
89259-886EA 169.23 MXN
89259-886 89259-890 89259-888
VWR® Micro Spatulas, Double-Ended
Espátulas
These VWR® Micro spatulas are double-ended and stainless steel.
  • Stainless steel construction
  • Double-ended design
Spatula has a curved spatula. Spatula is sharpened at the tip for cutting and separating and has a small knob at other end for crushing crystals.