VWR® Carboys with Versatile Caps, Dark Amber PP
Proveedor: VWR
15N-1111 15N-3121
89233-024EA 3471.55 MXN
89233-024 89233-032
VWR® Carboys with Versatile Caps, Dark Amber PP
Frascos Garrafones
VWR® dark amber carboys are designed to protect photo-sensitive materials, while providing users with a highly versatile container that maximizes storage efficiency and ease of use.
  • Polypropylene fabrication
  • Autoclavable
  • Optional wide neck enables easier cleaning
  • Secure lifting with molded grips
The distinctive rectangular shape saves valuable bench space.
Made in the USA.
Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows the user to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the container as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy. dark amber carboys are made from polypropylene, in compliance with FDA requirements for food use. Versatile caps are constructed of polypropylene and are autoclavable at 121 °C (250 °F) for fifteen minutes.