VWR® Safety Cabinets for Flammables and Corrosives
Proveedor: Avantor
VWR®
826022 829080 823022 823000 823080 826020 824520 824580 829000 829020 823020 826002 826080 824522 823002 826000 824500 824502
89522-648EA 36122.33 MXN
89522-618 89522-648 89522-626 89522-636 89522-628 89522-638 89522-616 89522-622 89522-644 89522-632 89522-624 89522-646 89522-634 89522-640 89522-650 89522-620 89522-642 89522-630
VWR® Safety Cabinets for Flammables and Corrosives
Mobiliario Armarios de almacenamiento
VWR® Safety Cabinets are designed to protect both laboratory personnel and the cabinet contents
  • Fully welded, 18-gauge double-wall construction for fire protection
  • Upper and lower diametrically opposed side vents
  • All models fire tested and approved by FM Global
Fully welded, 18-gauge galvanized steel ensures fire protection and strength with reduced gaps. Equipped with upper and lower diametrically opposed side vents, a ground connector on lower right side of the cabinet, double-wall construction with 3.8 cm (11/2”) insulating air space, and an environmentally friendly, electrostatically applied powder coat finish.
Cabinets are available for either flammables (yellow) or corrosives (blue).
Certificaciones: All door styles meet OSHA 1910.106, NFPA 30 and NFPA 1. Self-close door styles also meet IFC requirements and feature a fusible link that melts at 74°C (165°F) for automatic closure in the event of a fire.