Koolit® Foam Brick Gel Packs, Gel Pack Refrigerants, Cold Chain Technologies
Proveedor: Cold Chain Technologies
Koolit®
313F 306F 301F 305F 304F 309F
33500-338CS 863.43 MXN
66025-722 33500-336 66025-796 33500-334 33500-338 33500-340
Koolit® Foam Brick Gel Packs, Gel Pack Refrigerants, Cold Chain Technologies
Envases fríos
Formulated from nontoxic materials, these reusable foam brick gel packs maintain the thermal profile of temperature-sensitive products safely and more efficiently than ice.
The puncture-resistant foam brick gel packs retain a uniform shape and are ideal for exact or tight-fitting configurations. Even heat absorption rate allows for greater cooling longevity.