Koolit® Gel Bottles, Cold Chain Technologies
Proveedor: Cold Chain Technologies
Koolit®
404 405 406
33500-344CS 2883.02 MXN
33500-344 33500-342 33500-346
Koolit® Gel Bottles, Cold Chain Technologies
Envases fríos
Heavy-duty, tough and puncture-proof, our bottles are ideal for rough environments and reuse. Koolit bottles are made of blow-molded high-density plastic that holds 0 ºC (32 ºF) gel formulation.
  • Dependable, durable and heavy-duty, re-usable, protection for refrigerated and frozen shipments
  • Tough and puncture proof bottles are ideal for rough environments and reuse
  • High-quality, non-toxic materials that meet or exceed the standards of USDA, HAACP, DOT, and all major carriers, including FedEx® and UPS®
Available in three sizes and weights