Trays, Dispensing, High Density Polyethylene, Dynalon
Proveedor: Dynalon
108884-8 108884-1 108884-4 108884-6
62661-914CS 2850.31 MXN
30620-518 62661-900 62661-914 62661-904
Trays, Dispensing, High Density Polyethylene, Dynalon
Bandejas/Platos/Ollas Bandejas/Placas de laboratorio
Heavy trays fabricated with 1/4" thick high density polyethylene
All welded construction. Dispensing trays are supplied with polypropylene dispensing valve located as close to the tray bottom as possible (can be in other locations upon request). All trays can be customized with spigots and covers which are fabricated to order
Información para pedidos: Additional sizes and materials are available upon request.