Ward's® Single Probes Base Unit w/Ambient Temperature Sensor
Proveedor: EISCO SCIENTIFIC LLC
WSB-101-500
470017-388EA 6043.25 MXN
470017-388
Ward's® Single Probes Base Unit w/Ambient Temperature Sensor
Pruebas de temperatura
Just one Ward's Single Probes Base Unit is all you need to start collecting data quickly and easily with any Ward's Single Probes Sensor.
  • Built-in sensor
  • Display data in real time
  • Universal connectivity
Featuring a built-in ambient temperature sensor, this versatile base unit can be used on its own to measure and record temperature data, or connected interchangeably to any Ward's Single Probes Sensor to automatically engage the unit and measure and display data in real time. Whether your classroom is equipped with a PC, Mac, iPad, Android tablet, Smartphone, or any combination of these, the Ward's Single Probes Base Unit offers universal connectivity no matter which device you choose to use. Data can be displayed directly on the easy-to-read touch screen built into the unit, transmitted to your PC/Mac via USB cable, or wirelessly to any PC/Mac, tablet, or Smartphone using the Ward's Single Probes Wi-Fi Module (available separately). Student-friendly icon-based touch screen is easy for students to understand and use. Select from four different data display modes to see data in digital, analog, graph, or barograph format. Includes user guide and USB cable. Software download available online. Ward's Single Probes Sensors and Wi-Fi Module available separately.