Neogard® Powder-Free Chloroprene Examination Gloves, Microflex®
Proveedor: Ansell
C524 C523 C521 C520 C522 C522 C521 C523 C524
10159-880BX 499.97 MXN
CA10837-056 CA10118-980 10159-880 CA10159-880 10118-980 CA10159-878 CA10160-604 10160-604 10159-878
Neogard® Powder-Free Chloroprene Examination Gloves, Microflex®
Guantes
The Neogard glove is made from a unique, non latex formulation that delivers outstanding grip in wet and dry environments.
  • Ambidextrous
  • Beaded cuffs
  • Textured fingers
With unparalleled tactile sensitivity and strength, Neogard gloves are ideal for every task. Textured fingers ensure a strong and secure grip, while the advanced formulation helps deliver excellent wet and dry grip.