Nylon Mesh Cell Strainers
15-1070
490005-981CS 3700 MXN
490005-981
Nylon Mesh Cell Strainers
Tamices celulares
Nylon Mesh and compatible with 50 mL Centrifuge Tubes. Features of a polypropylene frame with a molded tab for easy handling. Gamma radiation sterilized, non-pyrogenic, and individually wrapped.
  • Compatible with 50 mL centrifuge tubes
  • Polypropylene frame with molded handling tab
  • Gamma radiation sterilized
  • Non-pryogenic
  • Individually wrapped
  • Sterile