VEE GEE VanGuard® Stereo Microscopes, 1200ZP Series
1275ZP
490011-024EA 25191.77 MXN
490011-024
VEE GEE VanGuard® Stereo Microscopes, 1200ZP Series
VanGuard 1200ZP Series Stereo Microscopes feature precision stereo optics producing the ultimate in high-resolution, three-dimensional images
All models feature 10X Wide Field eyepieces, spring-loaded stage clips, pole stands, and binocular heads.
Optional eyepieces (15X & 20X) and supplementary lenses are available customize any model for specific magnification requirements.
Información para pedidos: Microscopes come with a five year, limited warranty.