Dynalon Cuvette Stirring Spatulas
Proveedor: Dynalon
375985
490007-182CS 1804.32 MXN
490007-182
Dynalon Cuvette Stirring Spatulas
Imánes de agitación Varillas agitadoras
Polystyrene disposable Cuvette Stirring Spatulas measure 120 mm long by 3 mm diameter
Packed 500 stirrers in a polyethylene bag, 2 bags (1000 stirrers) per case.