VWR® Ultrasonic Cleaners, 220 V
Proveedor: Avantor
SK8200B SK5200B SK5200BT SK3300BT SK8200BT SK2200B SK8300B SK3300GT SK8300GT SK2200GT SK8300BT SK7200GT SK1200GT SK1200B SK1200BT SK7200B SK5200GT SK2200BT SK3300B
97043-938EA 29442.39 MXN
97044-012 97044-000 97043-958 97043-938 97044-008 97043-994 97043-962 97043-930 97043-974 97043-986 97043-942 97043-954 97043-966 97043-978 97043-934 97043-946 97043-990 97043-970 97043-982
VWR® Ultrasonic Cleaners, 220 V
Baños ultrasónicos
Ultrasonic cleaners provide highly effective and efficient cleaning of laboratory glassware and instruments.
  • Highly effective and efficient cleaning
  • Digital timer
  • Optional features and accessories available
All models feature a digital timer. Optional features include a heater and degassing function. Accessory baskets, covers, and support racks are constructed of Type 304 stainless steel.
Información para pedidos: Accessories are available; contact your sales representative for more information.