Jenway 73 Series Spectrophotometer, Cole Parmer, Antylia scientific
Proveedor: Cole Parmer
83058-18 83058-15
89168-830 89168-836
Jenway 73 Series Spectrophotometer, Cole Parmer, Antylia scientific
Espectrofotómetros y Fotómetros Espectrofotómetros UV/Vis
The 73 series introduces four new spectrophotometers to the extensive Jenway range
All of the models in the 73 series range are covered by a three year warranty which includes the xenon lamp. These spectrophotometers are easy to use and ideal for use in education and general QC laboratories. 
The advanced models have the additional feature of an easy access USB port on the front of the unit which enables results to be stored directly to a USB memory stick for easy transfer of data. These models can also store up to 240 methods which can be copied from one unit to another using the USB capability.
These spectrophotometers use icon driven software and have an improved navigation system for easy and intuitive usability. The range includes two basic models, 7300 and 7305 and two advanced models, 7310 and 7315. The 7300 and 7305 feature measurement modes for absorbance, % transmittance and concentration. 