Cobalt® Powder-Free Nitrile Examination Gloves, Microflex®
Proveedor: Ansell
N192 N191 N193 N193 N194 N194 N190 N190 N191 N192
89500-618CS 2872.8 MXN
CA75841-204 75841-204 89500-618 CA89500-620 CA89500-622 89500-624 CA89500-624 89500-622 89500-620 CA89500-618
Cobalt® Powder-Free Nitrile Examination Gloves, Microflex®
Guantes
This soft and stretchy nitrile glove is the best-selling glove from the Microflex® brand of products.
  • Ambidextrous
  • Nonsterile
  • Beaded cuffs
  • Exceptional tactile sensitivity
  • Powder-free
  • Durable and resistant to punctures
Durable and puncture resistant with a fully textured surface for enhanced grip. Ideal for jobs requiring exceptional tactile sensitivity. Gloves are 'cobalt' blue, nonsterile, and ambidextrous. They are also fully textured and feature beaded cuffs.