Eppendorf 5427R Microcentrifuges
: Avantor
Labs demanding high sample throughput will benefit from the 5427R, featuring a double-row 48-place rotor to increase efficiency and improve running time.
- 48 x 1.5/2.0 mL capacity rotor available
- Eppendorf QuickLock lids come standard with all aerosol-tight rotors
- Automatic rotor recognition and imbalance detection for maximum safety
- Maintains constant 4°C at maximum speed
- FastTemp program is optimized for super-fast 11-minute pre-cooling and maximum temperature accuracy inside the rotor
The 5427R provides refrigeration with innovative compressor technology that continuously cools even after the run, so your samples stay safe.