VWR® Pipette Tips, Extra Long
Proveedor: VWR
VWR®
1037-800-300
89368-966CS 5703.84 MXN
89368-966
VWR® Pipette Tips, Extra Long
Puntas de pipetas
These polypropylene tips are longer than most 10 µl tips to allow users to reach down into 0.65 ml tubes and multiwell plates.
  • RNase/DNase-, endotoxin-, protease-free, ATP-free, BPA -fee
  • Tips and racks are autoclavable
  • Supplied in bags or refillable racks
Tips feature a fine thin capillary section at the end that is non bevelled and very flexible; enables users to dispense the last small amount of sample from the tip.