Cole-Parmer® BS-200 Series Stirrers for Cell Culture Stirring Systems, Antylia Scientific
Proveedor: Antylia Scientific
21837-02 21837-03 21837-04 21837-01
35756-422EA 65754.59 MXN
35756-418 35756-416 35756-422 35756-420
Cole-Parmer® BS-200 Series Stirrers for Cell Culture Stirring Systems, Antylia Scientific
Imánes de agitación Agitadores magnéticos multiposición
Gentle stirring action promotes high cell yields.
  • Soft start/stop avoids excessive turbulence in the culture media
  • Interval stirring is ideal for extremely fragile cells
The cell culture stirring system incorporates everything needed to achieve high cell yields. The soft start/stop feature provides slow acceleration and deceleration of the stirrer, avoiding excessive turbulence in the culture media and eliminating cell damage. Use the interval stirring mode during the attachment phase to further reduce agitation of the media or when culturing particularly fragile cells. The stirrers create virtually no heat making them suitable for use in both incubators and cold rooms.
Use the biological stirrer with Cole-Parmer® Essentials culturing flasks - order separately. The culture flasks with a unique base design and the bulb-ended stirrer ensure that the cells are lifted into suspension at the lowest possible speeds promoting high cell yields.
Información de suministro: Supplied with three power cords with plug (US, UK, and European).