BOD Measurement, OxiTop® IS12 - 6
Proveedor: WTW
208212
80085-460
BOD Measurement, OxiTop® IS12 - 6
DBO, Demanda biológica de oxígeno
BOD respirometric measurement without mercury (Hg) according to EN 1889-2 for auto-control
The OxiTop® system was designed for the measurement of BOD from wastewater samples that are not highly contaminated, by measurement of the decrease of oxygen pressure and absorption of CO₂ generated in the sample bottle by NaOH.
To be used in conjunction with a magnetic stirrer platform and a thermostatic cabinet.
IS12-6 set is a ready to use package for 6 simultaneous measurements (extendable to 12) with an IS Inductive stirring system for 12 places, and 6 OxiTop® heads and accessories as above.
  • Very simple to use and mercury-free
  • Recorded values can be memorised for up to five days or viewed instantaneously on the numerical display
  • 6 and 12 sample bottle versions available
  • Measurement ranges: 0...40/80/200/400/800/2000/4000 mg/l BOD
  • Auto Temp function - if the sample is too cold, the start of the measurement is delayed
Información de suministro: The IS sets include the ready to use package of sample bottles, stirrer platform and stirrer bars, stirrer bar retriever, rubber sleeves, NaOH pellets, 1 each of overflow measuring flask 164 ml or 432 ml, nitrification inhibitor (thermostatic cabinet not included).