Portable Photometers, pHotoFlex, WTW
Proveedor: WTW
pHotoFlex®
251210
97054-112EA 2745.6 MXN
97054-112
Portable Photometers, pHotoFlex, WTW
Espectrofotómetros y Fotómetros Espectrofotómetros UV/Vis
A practical and robust series of portable photometers, suitable for a wide range of photometry, turbidity and pH applications in the areas of environmental monitoring, water treatment, wine and beverage industries and process control.The pHotoFLex® series offers more than 120 methods for commercial test kits and option for 100 user-defined programmes. Other features include a backlit display with automatic switch off. The PhotoFlex STD has 160 methods for commercial test kits with 10 user-defined programmes.
  • Able to accommodate 16 mm (variable height) and 28 mm cuvettes by means of a fixed adapter
  • User guidance via display for easy operation without handbook
  • Integrated pH measurement with automatic temperature compensation (except STD model)
  • Turbidity measurement with infrared light source according to DIN 27027/ISO 7027 for 665-0169
  • Battery operation with four AA batteries for approximately 3000 measurements
pH measuring range - 2.00 ... + 16.00. Resolution 0.01
Información de suministro: The pHotoFlex®/Set includes the pHotoFlex portable photometer with pH in a field case with tray to hold instruments and accessories.
The pHotoFlex® Turb/SET includes the pHotoFlex portable photometer with integrated turbidity and pH function in a field case with appropriate accessories.