Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
Login
/
Register
Mexico
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
$0.00
Order Entry
Home
Brushes
Vikan® Hand Scrub Brushes, Stiff, Remco Products
Vikan® Hand Scrub Brushes, Stiff, Remco Products
Vikan® Hand Scrub Brushes, Stiff, Remco Products
76007-388
:
Avantor
Expand image
Vikan® Hand Scrub Brushes, Stiff, Remco Products
76007-388
:
Avantor
Bristle Color:
Blue
Bristle Material:
Polyester
Handle Material:
Plastic
Total length:
9.8"
Cat. no.:
76007-388
Handle Color:
Blue
Width:
4.75"