Qualitative Filter Paper, Grade 601, Ahlstrom
Proveedor: Ahlstrom
6010-1500 6010-2700 6010-3850 6010-1100 6010-3200 6010-0300 6010-1250 6010-0750 6010-0250 6010-0900 6010-2500 6010-4000 6010-1850 6010-0550 6010-0700 6010-0470 6010-2400 6010-0425 6010-5000
47745-814PK 250.8 MXN
38000-025 47745-810 38000-229 47749-670 47749-662 47749-660 47749-666 47749-656 47749-664 47745-808 47749-668 47749-658 38000-160 38000-182 47745-814 47745-812 28342-016 47745-818 47745-816
Qualitative Filter Paper, Grade 601, Ahlstrom
Filtros Papeles de filtros Papel de filtro cualitativo
Ahlstrom Grade 601 is used in a wide range of laboratory applications such as clarifying liquids, rapid filtration of coarser precipitates, soil analysis, seed testing and more.
This low ash grade contains more than 95% super refined alpha-cellulose and linter fibers.