VWR® Glass Filter Holders
Proveedor: VWR
VWR®
262.202.01 260.202.01
89428-962CS 54056.22 MXN
89428-962 89428-952
VWR® Glass Filter Holders
Portafiltros
Glass components of this unit are manufactured from borosilicate 3.3 glass.
  • Borosilicate glass funnel and base
  • Anodized aluminum spring clamp
  • Silicone stopper
No. 8 perforated silicone stopper mounts in standard 1 l and 4 l filtering flasks.
Información de suministro: Supplied with a coarse porosity fritted or Stainless Steel support base, 300 ml graduated funnel, anodized aluminum clamp, and a no. 8 silicone stopper.