VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialized
Proveedor: VWR
VWR®
131.236.08 131.236.01 131.236.02 131.236.05 131.236.07 131.236.04 131.236.06 131.236.03
10124-074CS 15996.1 MXN
10124-074 10124-084 10124-072 10124-170 10124-070 10124-168 10124-398 10124-166
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialized
Matraces Matraces aforados
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
  • Borosilicate glass fabrication
  • Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with glass penny head stopper.
Información para pedidos: Flasks come with individual certificate of compliance.